missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Modulator of apoptosis 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Modulator of apoptosis 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6372-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_071434.2 | |
Modulator of apoptosis 1 Protein | |
Research Use Only | |
MOAP1 | |
Recombinant Protein | |
E. coli | |
MTLRLLEDWC RGMDMNPRKA LLIAGISQSC SVAEIEEALQ AGLAPLGEYR LLGRMFRRDE NRKVALVGLT AETSHALVPK EIPGKGGIWR VIFKPPDPDN TFLSRLNEFL AGEGMTVGEL SRALGHENGS LDPEQGMIPE MWAPMLAQAL EALQPALQCL KYKKLRVFSG RESPEPGEEE FGRWMFHTTQ MIKAWQVPDV EKRRRLLESL RGPALDVIRV LKINNPLITV DECLQALEEV FGVTDNPREL QVKYLTTYQK DEEKLSAYVL RLEPLLQKLV QRGAIERDAV NQARLDQVIA GAVHKTIRRE LNLPEDGPAP GFLQLLVLIK DYEAAEEEEA LLQAILEGNF T | |
Greater than 80% as determined by SDS-PAGE. |
64112 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |