missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP7 Protein
A DNA sequence encoding the Homo sapiens (Human) MMP7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8575-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002414.1 | |
MMP7 Protein | |
Research Use Only | |
MMP7 | |
Recombinant Protein | |
E. coli | |
YSLFPNSPKW TSKVVTYRIV SYTRDLPHIT VDRLVSKALN MWGKEIPLHF RKVVWGTADI MIGFARGAHG DSYPFDGPGN TLAHAFAPGT GLGGDAHFDE DERWTDGSSL GINFLYAATH ELGHSLGMGH SSDPNAVMYP TYGNGDPQNF KLSQDDIKGI QKLYGKRSNS RKK | |
Greater than 90% as determined by SDS-PAGE. |
4316 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |