missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MKI67 Protein

A DNA sequence encoding the Homo sapiens (Human) MKI67, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6362-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022760

  • 24050.00 NOK / 1mg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001139438.1
MKI67 Protein
Research Use Only
MKI67
Recombinant Protein
E. coli
RKLDAEDVIG SRRQPRAPKE KAQPLEDLAS FQELSQTPGH TEELANGAAD SFTSAPKQTP DSGKPLKISR RVLRAPKVEP VGDVVSTRDP VKSQSKSNTS LPPLPFKRGG GKDGSVTGTK RLRCMPAPEE IVEELPASKK QRVAPRARGK SSEPVVIMKR SLRTSAKRIE PAEELNSNDM KTNKEEHKLQ DSVPENKGIS LRSRRQNKTE AEQQITEVFV LAERIEINRN EKKPMKTSPE MDIQNPDDGA RKPIPRDKVT ENKRCLRSAR QNESSQPKVA EESGGQKSAK VLMQNQKGKG EAGNSDSMCL RSRKTKSQPA ASTLESKSVQ RVTRSVKRCA ENPKKAEDNV CVKKIRTRSH RDSEDI
Greater than 90% as determined by SDS-PAGE.
4288
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers