missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MKI67 Protein
A DNA sequence encoding the Homo sapiens (Human) MKI67, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6362-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001139438.1 | |
MKI67 Protein | |
Research Use Only | |
MKI67 | |
Recombinant Protein | |
E. coli | |
RKLDAEDVIG SRRQPRAPKE KAQPLEDLAS FQELSQTPGH TEELANGAAD SFTSAPKQTP DSGKPLKISR RVLRAPKVEP VGDVVSTRDP VKSQSKSNTS LPPLPFKRGG GKDGSVTGTK RLRCMPAPEE IVEELPASKK QRVAPRARGK SSEPVVIMKR SLRTSAKRIE PAEELNSNDM KTNKEEHKLQ DSVPENKGIS LRSRRQNKTE AEQQITEVFV LAERIEINRN EKKPMKTSPE MDIQNPDDGA RKPIPRDKVT ENKRCLRSAR QNESSQPKVA EESGGQKSAK VLMQNQKGKG EAGNSDSMCL RSRKTKSQPA ASTLESKSVQ RVTRSVKRCA ENPKKAEDNV CVKKIRTRSH RDSEDI | |
Greater than 90% as determined by SDS-PAGE. |
4288 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |