missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MCP-3 / CCL7 Protein
A DNA sequence encoding the Homo sapiens (Human) MCP-3 / CCL7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8526-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006264.2 | |
MCP-3 / CCL7 Protein | |
Research Use Only | |
CCL7 | |
Recombinant Protein | |
E. coli | |
QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL | |
Greater than 90% as determined by SDS-PAGE. |
6354 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |