missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAP2K3 Protein
A DNA sequence encoding the Homo sapiens (Human) MAP2K3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6336-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001303261.1 | |
MAP2K3 Protein | |
Research Use Only | |
MAP2K3 | |
Recombinant Protein | |
E. coli | |
MESPASSQPA SMPQSKGKSK RKKDLRISCM SKPPAPNPTP PRNLDSRTFI TIGDRNFEVE ADDLVTISEL GRGAYGVVEK VRHAQSGTIM AVKRIRATVN SQEQKRLLMD LDINMRTVDC FYTVTFYGAL FREGDVWICM ELMDTSLDKF YRKVLDKNMT IPEDILGEIA VSIVRALEHL HSKLSVIHRD VKPSNVLINK EGHVKMCDFG ISGYLVDSVA KTMDAGCKPY MAPERINPEL NQKGYNVKSD VWSLGITMIE MAILRFPYES WGTPFQQLKQ VVEEPSPQLP ADRFSPEFVD FTAQCLRKNP AERMSYLELM EHPFFTLHKT KKTDIAAFVK EILGEDS | |
Greater than 90% as determined by SDS-PAGE. |
5606 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |