missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Major capsid protein L1 Protein
A DNA sequence encoding the Homo sapiens (Human) Major capsid protein L1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8600-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_040317.1 | |
Major capsid protein L1 Protein | |
Research Use Only | |
L1 | |
Recombinant Protein | |
E. coli | |
GSIVTSDSQL FNKPYWLHKA QGHNNGVCWH NQLFVTVVDT TPSTNLTICA STQSPVPGQY DATKFKQYSR HVEEYDLQFI FQLCTITLTA DVMSYIHSMN SSILEDWNFG VPPPPTTSLV DTYRFVQSVA ITCQKDAAPA ENKDPYDKLK FWNVDLKEKF SLDLDQYPLG RKFLVQ | |
Greater than 90% as determined by SDS-PAGE. |
1489090 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |