missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LY6G6D Protein

A DNA sequence encoding the Homo sapiens (Human) LY6G6D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9258-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077931

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_067069.2
LY6G6D Protein
Research Use Only
LY6G6D
Recombinant Protein
Yeast
NRMRCYNCGG SPSSSCKEAV TTCGEGRPQP GLEQIKLPGN PPVTLIHQHP ACVAAHHCNQ VETESVGDVT YPAHRDCYLG DLCNS
Greater than 90% as determined by SDS-PAGE.
58530
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers