Learn More
enQuireBio™ Recombinant Human LY6G6D Protein
A DNA sequence encoding the Homo sapiens (Human) LY6G6D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9258-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_067069.2 | |
LY6G6D Protein | |
Research Use Only | |
LY6G6D | |
Recombinant Protein | |
Yeast | |
NRMRCYNCGG SPSSSCKEAV TTCGEGRPQP GLEQIKLPGN PPVTLIHQHP ACVAAHHCNQ VETESVGDVT YPAHRDCYLG DLCNS | |
Greater than 90% as determined by SDS-PAGE. |
58530 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |