missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ILDR2 Protein

A DNA sequence encoding the Homo sapiens (Human) ILDR2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7756-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16080621

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_955383.1
ILDR2 Protein
Research Use Only
ILDR2
Recombinant Protein
Yeast
MDRVLLRWIS LFWLTAMVEG LQVTVPDKKK VAMLFQPTVL RCHFSTSSHQ PAVVQWKFKS YCQDRMGESL GMSSTRAQSL SKRNLEWDPY LDCLDSRRTV RVVASKQGST VTLGDFYRGR EITIVHDADL QIGKLMWGDS GLYYCIITTP DDLEGKNEDS VELLVLGRTG LLADLLPSFA VEIMPE
Greater than 90% as determined by SDS-PAGE.
387597
50 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers