missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-21 Protein

Product Code. p-7083338
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Product Code. Quantity unitSize
18127760 10 μg 10µg
18749247 50 μg 50µg
2 options
This item is not returnable. View return policy

Product Code. 18127760

Brand: R&D Systems™ 8879IL010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-21 Protein is derived from E. coli. The Recombinant Human IL-21 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HBE4
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 59067
Molecular Weight (g/mol) Observed M.W.: 17kDa, reducing conditions, Theoretical M.W.: 15kDa
Quantity 10 μg
Source E. coli-derived human IL-21 protein Gln25-Ser155,with a N-terminal Met
Storage Requirements 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform
Common Name IL-21
Gene Symbol IL21
Biological Activity Measured in a cell proliferation assay using N1186 human T cells. Parrish-Novak, J. et al. (2000) Nature 408:57. The ED50 for this effect is typically 2-12ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Gln25-Ser155, with a N-terminal Met
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.