missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-21 Protein
Description
The Recombinant Human IL-21 Protein is derived from E. coli. The Recombinant Human IL-21 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q9HBE4 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 59067 |
| Molecular Weight (g/mol) | Observed M.W.: 17kDa, reducing conditions, Theoretical M.W.: 15kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human IL-21 protein Gln25-Ser155,with a N-terminal Met |
| Storage Requirements | 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?