missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF1R / CD221 Protein
A DNA sequence encoding the Homo sapiens (Human) IGF1R / CD221, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6207-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000866.1 | |
IGF1R / CD221 Protein | |
Research Use Only | |
IGF1R | |
Recombinant Protein | |
E. coli | |
YNITDPEELE TEYPFFESRV DNKERTVISN LRPFTLYRID IHSCNHEAEK LGCSASNFVF ARTMPAEGAD DIPGPVTWEP RPENSIFLKW PEPENPNGLI LMYEIKYGSQ VEDQRECVSR QEYRKYGGAK LNRLNPGNYT ARIQATSLSG NGSWTDPVFF YVQAKTGYE | |
Greater than 90% as determined by SDS-PAGE. |
3480 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |