missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNAR2 / IFNABR Protein
A DNA sequence encoding the Homo sapiens (Human) IFNAR2 / IFNABR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6204-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000865.2 | |
IFNAR2 / IFNABR Protein | |
Research Use Only | |
Ifnar2 | |
Recombinant Protein | |
E. coli | |
ISYDSPDYTD ESCTFKISLR NFRSILSWEL KNHSIVPTHY TLLYTIMSKP EDLKVVKNCA NTTRSFCDLT DEWRSTHEAY VTVLEGFSGN TTLFSCSHNF WLAIDMSFEP PEFEIVGFTN HINVMVKFPS IVEEELQFDL SLVIEEQSEG IVKKHKPEIK GNMSGNFTYI IDKLIPNTNY CVSVYLEHSD EQAVIKSPLK CTLLPPGQES ESAESAK | |
Greater than 90% as determined by SDS-PAGE. |
3455 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |