missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DMA Protein

A DNA sequence encoding the Homo sapiens (Human) HLA-DMA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7191-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017630

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_006111.2
HLA-DMA Protein
Research Use Only
HLA-DMA
Recombinant Protein
E. coli
VPEAPTPMWP DDLQNHTFLH TVYCQDGSPS VGLSEAYDED QLFFFDFSQN TRVPRLPEFA DWAQEQGDAP AILFDKEFCE WMIQQIGPKL DGKIPVSRGF PIAEVFTLKP LEFGKPNTLV CFVSNLFPPM LTVNWQHHSV PVEGFGPTFV SAVDGLSFQA FSYLNFTPEP SDIFSCIVTH EIDRYTAIAY WVPRNALPSD LLENVLC
Greater than 90% as determined by SDS-PAGE.
3108
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers