Learn More
enQuireBio™ Recombinant Human Histone H2B type 1-C / E / F / G / I Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2B type 1-C/E/F/G/I, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8382-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003509.1 | |
Histone H2B type 1-C / E / F / G / I Protein | |
Research Use Only | |
H2BFL | |
Recombinant Protein | |
E. coli | |
PEPAKSAPAP KKGSKKAVTK AQKKDGKKRK RSRKESYSVY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSS | |
Greater than 90% as determined by SDS-PAGE. |
3017 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |