missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HGF A Protein

A cDNA sequence encoding the HGF A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12229-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929768

  • 2140.00 NOK / 10µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HGF A Protein
Research Use Only
Recombinant Protein
E. coli
QRKRRNTIHEFKKSAKTTLIKIDPALKIKTKKVNTADQCANRCTRNKGLPFTCKAFVFDKARKQCLWFPFNSMSSGVKKEFGHEFDLYENKDYIRNCIIGKGRSYKGTVSITKSGIKCQPWSSMIPHEHSFLPSSYRGKDLQENYCRNPRGEEGGPWCFTSNPEVRYEVCDIPQCSEVECMTCNGESYRGLMDHTESGKICQRWDHQTPHRHKFLPERYPDKGFDDNYCRNPDGQPRPWCYTLDPHTRWEYCAIKTCADNTMNDTDVPLETTECIQGQGEGYRGTVNTIWNGIPCQRWDSQYPHEHDMTPENFKCKDLRENYCRNPDGSESPWCFTTDPNIRVGYCSQIPNCDMSHGQDCYRGNGKNYMGNLSQTRSGLTCSMWDKNMEDLHRHIFWEPDASKLNENYCRNPDDDAHGPWCYTGNPLIPWDYCPISRCEGDTTPTIVNLDHPVISCAKTKQLR
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HGF-A protein is supplied in 25mM Na. Acetate pH4.8, 1mM EDTA and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers