missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heat shock protein beta-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Heat shock protein beta-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7544-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001532.1 | |
Heat shock protein beta-2 Protein | |
Research Use Only | |
HSPB2 | |
Recombinant Protein | |
E. coli | |
MSGRSVPHAH PATAEYEFAN PSRLGEQRFG EGLLPEEILT PTLYHGYYVR PRAAPAGEGS RAGASELRLS EGKFQAFLDV SHFTPDEVTV RTVDNLLEVS ARHPQRLDRH GFVSREFCRT YVLPADVDPW RVRAALSHDG ILNLEAPRGG RHLDTEVNEV YISLLPAPPD PEEEEEAAIV EP | |
Greater than 80% as determined by SDS-PAGE. |
3316 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |