missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heat shock 70 kDa protein 1
A DNA sequence encoding the Homo sapiens (Human) Heat shock 70 kDa protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8058-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002146.2 | |
Heat shock 70 kDa protein 1 | |
Research Use Only | |
HSPA6 | |
Recombinant Protein | |
E. coli | |
MQAPRELAVG IDLGTTYSCV GVFQQGRVEI LANDQGNRTT PSYVAFTDTE RLVGDAAKSQ AALNPHNTVF DAKRLIGRKF ADTTVQSDMK HWPFRVVSEG GKPKVRVCYR GEDKTFYPEE ISSMVLSKMK ETAEAYLGQP VKHAVITVPA YFNDSQRQAT KDAGAIAGLN VLRIINEPTA AAIAYGLDRR GAGERNVLIF DLGGGTFDVS VLSIDAGVFE VKATAGDTHL GGEDFDNRLV NHFMEEFRRK HGKDLSGNKR ALRRLRTACE RAKRTLSSST QATLEIDSLF EGVDFYTSIT RARFEELCSD LFRSTLEPVE KALRDAKLDK AQIHDVVLVG GSTRIPKVQK LLQDFFNGKE LNKSINPDEA VAYGAAVQAA VLMGDKCEKV QDLLLLDVAP LSLGLETAGG VMTTLIQRNA TIPTKQTQTF TTYSDNQPGV FIQVYEGERA MTKDNNLLGR FELSGIPPAP RGVPQIEVTF DIDANGILSV TATDRSTGKA NKITITNDKG RLSKEEVERM VHEAEQYKAE DEAQRDRVAA KNSLEAHVFH VKGSLQEESL RDKIPEEDRR KMQDKCREVL AWLEHNQLAE KEEYEHQKRE LEQICRPIFS RLYGGPGVPG GSSCGTQARQ GDPSTGP | |
Greater than 90% as determined by SDS-PAGE. |
3310 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |