missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Granzyme H Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Human Granzyme H Protein is derived from NS0. The Recombinant Human Granzyme H Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl and Brij-35. |
| Gene ID (Entrez) | 2999 |
| Molecular Weight (g/mol) | 27 kDa |
| Name | Granzyme H |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Granzyme H protein Glu19-Leu246,with a C-terminal 10-His tag Accession &Num; P20718 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Structural Form | Pro form |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction