missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNB2 Protein
A DNA sequence encoding the Homo sapiens (Human) GNB2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8490-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005264.2 | |
GNB2 Protein | |
Research Use Only | |
GNB2 | |
Recombinant Protein | |
E. coli | |
ARKACGDSTL TQITAGLDPV GRIQMRTRRT LRGHLAKIYA MHWGTDSRLL VSASQDGKLI IWDSYTTNKV HAIPLRSSWV MTCAYAPSGN FVACGGLDNI CSIYSLKTRE GNVRVSRELP GHTGYLSCCR FLDDNQIITS SGDTTCALWD IETGQQTVGF AGHSGDVMSL SLAPDGRTFV SGACDASIKL WDVRDSMCRQ TFIGHESDIN AVAFFPNGYA FTTGSDDATC RLFDLRADQE LLMYSHDNII CGITSVAFSR SGRLLLAGYD DFNCNIWDAM KGDRAGVLAG HDNRVSCLGV TDDGMAV | |
Greater than 90% as determined by SDS-PAGE. |
2783 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |