missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutaminyl cyclase / QPCT Protein
A DNA sequence encoding the Homo sapiens (Human) Glutaminyl cyclase / QPCT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6564-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036545.1 | |
Glutaminyl cyclase / QPCT Protein | |
Research Use Only | |
QPCT | |
Recombinant Protein | |
E. coli | |
VSPSASAWPE EKNYHQPAIL NSSALRQIAE GTSISEMWQN DLQPLLIERY PGSPGSYAAR QHIMQRIQRL QADWVLEIDT FLSQTPYGYR SFSNIISTLN PTAKRHLVLA CHYDSKYFSH WNNRVFVGAT DSAVPCAMML ELARALDKKL LSLKTVSDSK PDLSLQLIFF DGEEAFLHWS PQDSLYGSRH LAAKMASTPH PPGARGTSQL HGMDLLVLLD LIGAPNPTFP NFFPNSARWF ERLQAIEHEL HELGLLKDHS LEGRYFQNYS YGGVIQDDHI PFLRRGVPVL HLIPSPFPEV WHTMDDNEEN LDESTIDNLN KILQVFVLEY LHL | |
Greater than 90% as determined by SDS-PAGE. |
25797 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |