missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Gamma-synuclein Protein
A DNA sequence encoding the Homo sapiens (Human) Gamma-synuclein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6707-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001317049.1 | |
Gamma-synuclein Protein | |
Research Use Only | |
SNCG | |
Recombinant Protein | |
E. coli | |
MDVFKKGFSI AKEGVVGAVE KTKQGVTEAA EKTKEGVMYV GAKTKENVVQ SVTSVAEKTK EQANAVSEAV VSSVNTVATK TVEEAENIAV TSGVVRKEDL RPSAPQQEGE ASKEKEEVAE EAQSGGD | |
Greater than 90% as determined by SDS-PAGE. |
6623 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |