missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galectin-9 Protein
A DNA sequence encoding the Homo sapiens (Human) Galectin-9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9244-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001317092.1 | |
Galectin-9 Protein | |
Research Use Only | |
LGALS9 | |
Recombinant Protein | |
Yeast | |
MAFSGSQAPY LSPAVPFSGT IQGGLQDGLQ ITVNGTVLSS SGTRFAVNFQ TGFSGNDIAF HFNPRFEDGG YVVCNTRQNG SWGPEERKTH MPFQKGMPFD LCFLVQSSDF KVMVNGILFV QYFHRVPFHR VDTISVNGSV QLSYISFQNP RTVPVQPAFS TVPFSQPVCF PPRPRGRRQK PPGVWPANPA PITQTVIHTV QSAPGQMFST PAIPPMMYPH PAYPMPFITT ILGGLYPSKS ILLSGTVLPS AQRFHINLCS GNHIAFHLNP RFDENAVVRN TQIDNSWGSE ERSLPRKMPF VRGQSFSVWI LCEAHCLKVA VDG | |
Greater than 90% as determined by SDS-PAGE. |
3965 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |