missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Galectin-1 / LGALS1 Protein

A DNA sequence encoding the Homo sapiens (Human) Galectin-1 / LGALS1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6294-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002296.1
Galectin-1 / LGALS1 Protein
Research Use Only
LGALS1
Recombinant Protein
E. coli
ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD
Greater than 90% as determined by SDS-PAGE.
3956
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers