missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GABARAP / Apg8p1 Protein
A DNA sequence encoding the Homo sapiens (Human) GABARAP / Apg8p1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8508-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009209.1 | |
GABARAP / Apg8p1 Protein | |
Research Use Only | |
GABARAP | |
Recombinant Protein | |
E. coli | |
KFVYKEEHPF EKRRSEGEKI RKKYPDRVPV IVEKAPKARI GDLDKKKYLV PSDLTVGQFY FLIRKRIHLR AEDALFFFVN NVIPPTSATM GQLYQEHHEE DFFLYIAYSD ESVYGL | |
Greater than 90% as determined by SDS-PAGE. |
11337 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |