missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Four and a half LIM domains protein 1

A DNA sequence encoding the Homo sapiens (Human) Four and a half LIM domains protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7560-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029390

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001230807.1
Four and a half LIM domains protein 1
Research Use Only
FHL3
Recombinant Protein
E. coli
SESFDCAKCN ESLYGRKYIQ TDSGPYCVPC YDNTFANTCA ECQQLIGHDS RELFYEDRHF HEGCFRCCRC QRSLADEPFT CQDSELLCND CYCSAFSSQC SACGETVMPG SRKLEYGGQT WHEHCFLCSG CEQPLGSRSF VPDKGAHYCV PCYENKFAPR CARCSKTLTQ GGVTYRDQPW HRECLVCTGC QTPLAGQQFT SRDEDPYCVA CFGELFAPKC SSCKRPIVGL GGGKYVSFED RHWHHNCFSC ARCSTSLVGQ GFVPDGDQVL CQGCSQAGP
Greater than 80% as determined by SDS-PAGE.
2275
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers