missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Four and a half LIM domains protein 1
A DNA sequence encoding the Homo sapiens (Human) Four and a half LIM domains protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7560-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001230807.1 | |
Four and a half LIM domains protein 1 | |
Research Use Only | |
FHL3 | |
Recombinant Protein | |
E. coli | |
SESFDCAKCN ESLYGRKYIQ TDSGPYCVPC YDNTFANTCA ECQQLIGHDS RELFYEDRHF HEGCFRCCRC QRSLADEPFT CQDSELLCND CYCSAFSSQC SACGETVMPG SRKLEYGGQT WHEHCFLCSG CEQPLGSRSF VPDKGAHYCV PCYENKFAPR CARCSKTLTQ GGVTYRDQPW HRECLVCTGC QTPLAGQQFT SRDEDPYCVA CFGELFAPKC SSCKRPIVGL GGGKYVSFED RHWHHNCFSC ARCSTSLVGQ GFVPDGDQVL CQGCSQAGP | |
Greater than 80% as determined by SDS-PAGE. |
2275 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |