missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human FGF-17 Protein

Product Code. 18614359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18614359 5 μg 5µg
18601618 500 μg 500µg
18650848 100 μg 100µg
18611738 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18614359 Supplier Novus Biologicals™ Supplier No. NBP2613125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

FGF-17 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine balb/c 3T3 cells is less than 10 ng/mL, corresponding to a specific activity of >1.0 x 10^5 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 8822
Molecular Weight (g/mol) MolecularWeight-theroretical: 22.6 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of FGF-17 AS determined by LAL method.
Gene Alias FGF-13, FGF-17, fibroblast growth factor 17
Gene Symbol FGF17
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Angiogenesis, Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein FGF-17
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.