missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM84B Protein
A cDNA sequence encoding the FAM84B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11850-20ug
Additional Details : Weight : 0.01000kg
Specifications
FAM84B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGNQVEK LTHLSYKEVP TADPTGVDRD DGPRIGVSYI FSNDDEDVEP QPPPQGPDGG GLPDGGDGPP PPQPQPYDPR LHEVECSVFY RDECIYQKSF APGSAALSTY TPENLLNKCK PGDLVEFVSQ AQYPHWAVYV GNFQVVHLHR LEVINSFLTD ASQGRRGRVV NDLYRYKPLS SSAVVRNALA HVGAKERELS WRNSESFAAW CRYGKREFKI GGELRIGKQP YRLQIQLSAQ RSHTLEFQSL EDLIMEKRRN DQIGRAAVLQ ELATHLHPAE PEEGDSNVAR TTPPPGRPPA PSSEEEDGEA VAH | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FAM84B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.15M NaCl, 1mM DTT and 0.1mM PMSF. |