missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Excitatory amino acid transporter 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Excitatory amino acid transporter 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6692-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001259016.1 | |
Excitatory amino acid transporter 4 Protein | |
Research Use Only | |
SLC1A6 | |
Recombinant Protein | |
E. coli | |
MVTIIHPGKG SKEGLHREGR IETIPTADAF MDLIRNMFPP NLVEACFKQF KTQYSTRVVT RTMVRTENGS EPGASMPPPF SVENGTSFLE NVTRALGTLQ EMLSFEETVP VPGSANGINA LGL | |
Greater than 90% as determined by SDS-PAGE. |
6511 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |