missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Excitatory amino acid transporter 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Excitatory amino acid transporter 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6692-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035160

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001259016.1
Excitatory amino acid transporter 4 Protein
Research Use Only
SLC1A6
Recombinant Protein
E. coli
MVTIIHPGKG SKEGLHREGR IETIPTADAF MDLIRNMFPP NLVEACFKQF KTQYSTRVVT RTMVRTENGS EPGASMPPPF SVENGTSFLE NVTRALGTLQ EMLSFEETVP VPGSANGINA LGL
Greater than 90% as determined by SDS-PAGE.
6511
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers