missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ETHE1 / HSCO Protein

A DNA sequence encoding the Homo sapiens (Human) ETHE1 / HSCO, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6004-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949289

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001307796.1
ETHE1 / HSCO Protein
Research Use Only
ETHE1
Recombinant Protein
E. coli
VARRQLSQRG GSGAPILLRQ MFEPVSCTFT YLLGDRESRE AVLIDPVLET APRDAQLIKE LGLRLLYAVN THCHADHITG SGLLRSLLPG CQSVISRLSG AQADLHIEDG DSIRFGRFAL ETRASPGHTP GCVTFVLNDH SMAFTGDALL IRGCGRTDFQ QGCAKTLYHS VHEKIFTLPG DCLIYPAHDY HGFTVSTVEE ERTLNPRLTL SCEEFVKIMG NLNLPKPQQI DFAVPANMRC GVQTPTA
Greater than 90% as determined by SDS-PAGE.
23474
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers