missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ERH / DROER Protein
A DNA sequence encoding the Homo sapiens (Human) ERH / DROER, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8423-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004441.1 | |
ERH / DROER Protein | |
Research Use Only | |
ERH | |
Recombinant Protein | |
E. coli | |
SHTILLVQPT KRPEGRTYAD YESVNECMEG VCKMYEEHLK RMNPNSPSIT YDISQLFDFI DDLADLSCLV YRADTQTYQP YNKDWIKEKI YVLLRRQAQQ AGK | |
Greater than 90% as determined by SDS-PAGE. |
2079 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |