missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ERH / DROER Protein

A DNA sequence encoding the Homo sapiens (Human) ERH / DROER, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8423-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084531

  • 5325.00 NOK / 100µg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004441.1
ERH / DROER Protein
Research Use Only
ERH
Recombinant Protein
E. coli
SHTILLVQPT KRPEGRTYAD YESVNECMEG VCKMYEEHLK RMNPNSPSIT YDISQLFDFI DDLADLSCLV YRADTQTYQP YNKDWIKEKI YVLLRRQAQQ AGK
Greater than 90% as determined by SDS-PAGE.
2079
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers