Learn More
R&D Systems™ Recombinant Human EN-RAGE Protein
Description
The Recombinant Human EN-RAGE Protein is derived from E. coli. The Recombinant Human EN-RAGE Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 6283 |
| Molecular Weight (g/mol) | 10.6 kDa |
| Name | EN-RAGE/S100A12 |
| Quantity | 50 μg |
| Source | E. coli-derived human EN-RAGE/S100A12 protein Met1-Glu92 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |