missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DTD2 Protein
A DNA sequence encoding the Homo sapiens (Human) DTD2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7941-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_542395.1 | |
DTD2 Protein | |
Research Use Only | |
DTD2 | |
Recombinant Protein | |
E. coli | |
MAEGSRIPQA RALLQQCLHA RLQIRPADGD VAAQWVEVQR GLVIYVCFFK GADKELLPKM VNTLLNVKLS ETENGKHVSI LDLPGNILII PQATLGGRLK GRNMQYHSNS GKEEGFELYS QFVTLCEKEV AANSKCAEAR VVVEHGTYGN RQVLKLDTNG PFTHLIEF | |
Greater than 90% as determined by SDS-PAGE. |
112487 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |