missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA oxidative demethylase ALKBH2 Protein

A DNA sequence encoding the Homo sapiens (Human) DNA oxidative demethylase ALKBH2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7771-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16090681

  • 5325.00 NOK / 100µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001001655.1
DNA oxidative demethylase ALKBH2 Protein
Research Use Only
ALKBH2
Recombinant Protein
E. coli
MDRFLVKGAQ GGLLRKQEEQ EPTGEEPAVL GGDKESTRKR PRREAPGNGG HSAGPSWRHI RAEGLDCSYT VLFGKAEADE IFQELEKEVE YFTGALARVQ VFGKWHSVPR KQATYGDAGL TYTFSGLTLS PKPWIPVLER IRDHVSGVTG QTFNFVLINR YKDGCDHIGE HRDDERELAP GSPIASVSFG ACRDFVFRHK DSRGKSPSRR VAVVRLPLAH GSLLMMNHPT NTHWYHSLPV RKKVLAPRVN LTFRKILLTK K
Greater than 80% as determined by SDS-PAGE.
121642
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers