missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA oxidative demethylase ALKBH2 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA oxidative demethylase ALKBH2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7771-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001655.1 | |
DNA oxidative demethylase ALKBH2 Protein | |
Research Use Only | |
ALKBH2 | |
Recombinant Protein | |
E. coli | |
MDRFLVKGAQ GGLLRKQEEQ EPTGEEPAVL GGDKESTRKR PRREAPGNGG HSAGPSWRHI RAEGLDCSYT VLFGKAEADE IFQELEKEVE YFTGALARVQ VFGKWHSVPR KQATYGDAGL TYTFSGLTLS PKPWIPVLER IRDHVSGVTG QTFNFVLINR YKDGCDHIGE HRDDERELAP GSPIASVSFG ACRDFVFRHK DSRGKSPSRR VAVVRLPLAH GSLLMMNHPT NTHWYHSLPV RKKVLAPRVN LTFRKILLTK K | |
Greater than 80% as determined by SDS-PAGE. |
121642 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |