missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA mismatch repair protein Msh6 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA mismatch repair protein Msh6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6388-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000170.1 | |
DNA mismatch repair protein Msh6 Protein | |
Research Use Only | |
MSH6 | |
Recombinant Protein | |
E. coli | |
MSRQSTLYSF FPKSPALSDA NKASARASRE GGRAAAAPGA SPSPGGDAAW SEAGPGPRPL ARSASPPKAK NLNGGLRRSV APAAPTSCDF SPGDLVWAKM EGYPWWPCLV YNHPFDGTFI REKGKSVRVH VQFFDDSPTR GWVSKRLLKP YTGSKSKEAQ KGGHFYSAKP EILRAMQRAD EALNKDKIKR LELAVCDEPS EPEEEEEMEV GTTYVTDKSE EDNEIESEEE VQPKTQGSRR SSRQIKKRRV ISDSESDIGG SDVEFKPDTK EEGSSDEISS GVGDSESEGL NSPVKVARKR KRMVTGNGSL KRKSSRKETP SATKQATSIS SETKNTLRAF SAPQNSESQA HVSGGGDDSS RPTVWYHETL EWLKEEKRRD EHRRRPDHPD FDASTLYVPE | |
Greater than 90% as determined by SDS-PAGE. |
2956 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |