missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA-binding protein inhibitor ID-2 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA-binding protein inhibitor ID-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6191-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002157.2 | |
DNA-binding protein inhibitor ID-2 Protein | |
Research Use Only | |
ID2 | |
Recombinant Protein | |
E. coli | |
MKAFSPVRSV RKNSLSDHSL GISRSKTPVD DPMSLLYNMN DCYSKLKELV PSIPQNKKVS KMEILQHVID YILDLQIALD SHPTIVSLHH QRPGQNQASR TPLTTLNTDI SILSLQASEF PSELMSNDSK ALCG | |
Greater than 90% as determined by SDS-PAGE. |
3398 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |