missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Delta-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Delta-like protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8179-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_058637.1
Delta-like protein 1
Research Use Only
DLL3
Recombinant Protein
E. coli
AGVFELQIHS FGPGPGPGAP RSPCSARLPC RLFFRVCLKP GLSEEAAESP CALGAALSAR GPVYTEQPGA PAPDLPLPDG LLQVPFRDAW PGTFSFIIET WREELGDQIG GPAWSLLARV AGRRRLAAGG PWARDIQRAG AWELRFSYRA RCEPPAVGTA CTRLCRPRSA PSRCGPGLRP CAPLEDECEA PLVCRAGCSP EHGFCEQPGE CRCLEGWTGP LCTVPVSTSS CLSPRGPSSA TTGCLVPGPG PCDGNPCANG GSCSETPRSF ECTCPRGFYG LRCEVSGVTC ADGPCFNGGL CVGGADPDSA YICHCPPGFQ GSNCEKRVDR CSLQPCRNGG LCLDLGHALR CRCRAGFAGP RCEHDLDDCA GRACANGGTC VEGGGAHRCS CALGFGGRDC RERADPCAAR PCAHGGRCYA HFSGLVCACA PGYMGARCEF PVHPDGASAL PAAPPGLRPG DPQRYL
Greater than 90% as determined by SDS-PAGE.
10683
10 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers