missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DDOST / OST48 Protein
A DNA sequence encoding the Homo sapiens (Human) DDOST / OST48, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8493-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002941.1 | |
DDOST / OST48 Protein | |
Research Use Only | |
DDOST | |
Recombinant Protein | |
E. coli | |
SHLAKVTAEV VLAHLGGGST SRATSFLLAL EPELEARLAH LGVQVKGEDE EENNLEVRET KIKGKSGRFF TVKLPVALDP GAKISVIVET VYTHVLHPYP TQITQSEKQF VVFEGNHYFY SPYPTKTQTM RVKLASRNVE SYTKLGNPTR SEDLLDYGPF RDVPAYSQDT FKVHYENNSP FLTITSMTRV IEVSHWGNIA VEENVDLKHT GAVLKGPFSR YDYQRQPDSG ISSIRSFKTI LPAAAQDVYY RDEIGNVSTS HLLILDDSVE MEIRPRFPLF GGWKTHYIVG YNLPSYEYLY NLGDQYALKM RFVDHVFDEQ VIDSLTVKII LPEGAKNIEI DSPYEISRAP DELHYTYLDT FGRPVIVAYK KNLVEQHIQD IVVH | |
Greater than 90% as determined by SDS-PAGE. |
6184 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |