missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome c1, heme protein, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome c1, heme protein, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8372-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001907.2 | |
Cytochrome c1, heme protein, mitochondrial Protein | |
Research Use Only | |
CYC1 | |
Recombinant Protein | |
E. coli | |
SDLELHPPSY PWSHRGLLSS LDHTSIRRGF QVYKQVCASC HSMDFVAYRH LVGVCYTEDE AKELAAEVEV QDGPNEDGEM FMRPGKLFDY FPKPYPNSEA ARAANNGALP PDLSYIVRAR HGGEDYVFSL LTGYCEPPTG VSLREGLYFN PYFPGQAIAM APPIYTDVLE FDDGTPATMS QIAKDVCTFL RWASEPEHDH RKRMGLKMLM MMALLVPLVY TIKRHKWSVL KSRKLAYRPP K | |
Greater than 90% as determined by SDS-PAGE. |
1537 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |