missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagenase 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Collagenase 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6367-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002418.1 | |
Collagenase 3 Protein | |
Research Use Only | |
MMP13 | |
Recombinant Protein | |
E. coli | |
YNVFPRTLKW SKMNLTYRIV NYTPDMTHSE VEKAFKKAFK VWSDVTPLNF TRLHDGIADI MISFGIKEHG DFYPFDGPSG LLAHAFPPGP NYGGDAHFDD DETWTSSSKG YNLFLVAAHE FGHSLGLDHS KDPGALMFPI YTYTGKSHFM LPDDDVQGIQ SLYGPGDEDP NPKHPKTPDK CDPSLSLDAI TSLRGETMIF KDRFFWRLHP QQVDAELFLT KSFWPELPNR IDAAYEHPSH DLIFIFRGRK FWALNGYDIL EGYPKKISEL GLPKEVKKIS AAVHFEDTGK TLLFSGNQVW RYDDTNHIMD KDYPRLIEED FPGIGDKVDA VYEKNGYIYF FNGPIQFEYS IWSNRIVRVM PANSILWC | |
Greater than 80% as determined by SDS-PAGE. |
4322 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |