Learn More
R&D Systems™ Recombinant Human Coagulation Factor Xa Protein
Product Code. 18389617
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389617 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18389617 Supplier R&D Systems™ Supplier No. 1063SE010
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
The Recombinant Human Coagulation Factor Xa Protein is derived from Sf 21 (baculovirus). The Recombinant Human Coagulation Factor Xa Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized from a 0.2 μm filtered solution in MES, NaCl and CaCl 2 .
Gene ID (Entrez) 2159
Molecular Weight (g/mol) 12 kDa & 30 kDa
Name Coagulation Factor X
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Coagulation Factor X protein Leu24-Lys488 with a C-terminal 10-His tag Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Active form
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 25 mM MES, 150 mM NaCl and 5 mM CaCl 2 pH 6.0.
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.