missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Charged multivesicular body protein 2a Protein
A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 2a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8478-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055268.1 | |
Charged multivesicular body protein 2a Protein | |
Research Use Only | |
BC2 | |
Recombinant Protein | |
E. coli | |
MDLLFGRRKT PEELLRQNQR ALNRAMRELD RERQKLETQE KKIIADIKKM AKQGQMDAVR IMAKDLVRTR RYVRKFVLMR ANIQAVSLKI QTLKSNNSMA QAMKGVTKAM GTMNRQLKLP QIQKIMMEFE RQAEIMDMKE EMMNDAIDDA MGDEEDEEES DAVVSQVLDE LGLSLTDELS NLPSTGGSLS VAAGGKKAEA AASALADADA DLEERLKNLR RD | |
Greater than 90% as determined by SDS-PAGE. |
27243 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |