missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Charged multivesicular body protein 1b Protein
A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 1b, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7751-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065145.2 | |
Charged multivesicular body protein 1b Protein | |
Research Use Only | |
CHMP1B | |
Recombinant Protein | |
E. coli | |
MSNMEKHLFN LKFAAKELSR SAKKCDKEEK AEKAKIKKAI QKGNMEVARI HAENAIRQKN QAVNFLRMSA RVDAVAARVQ TAVTMGKVTK SMAGVVKSMD ATLKTMNLEK ISALMDKFEH QFETLDVQTQ QMEDTMSSTT TLTTPQNQVD MLLQEMADEA GLDLNMELPQ GQTGSVGTSV ASAEQDELSQ RLARLRDQV | |
Greater than 80% as determined by SDS-PAGE. |
57132 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |