missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Charged multivesicular body protein 1
A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8158-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001182465.1 | |
Charged multivesicular body protein 1 | |
Research Use Only | |
CHMP5 | |
Recombinant Protein | |
E. coli | |
MNRLFGKAKP KAPPPSLTDC IGTVDSRAES IDKKISRLDA ELVKYKDQIK KMREGPAKNM VKQKALRVLK QKRMYEQQRD NLAQQSFNME QANYTIQSLK DTKTTVDAMK LGVKEMKKAY KQVKIDQIED LQDQLEDMME DANEIQEALS RSYGTPELDE DDLEAELDAL GDELLADEDS SYLDEAASAP AIPEGVPTDT KNKDGVLVDE FGLPQIPAS | |
Greater than 90% as determined by SDS-PAGE. |
51510 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |