Learn More
enQuireBio™ Recombinant Human CD112 / Nectin-2 / PVRL2 Protein
A DNA sequence encoding the Homo sapiens (Human) CD112 / Nectin-2 / PVRL2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7863-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001036189.1 | |
CD112 / Nectin-2 / PVRL2 Protein | |
Research Use Only | |
PVRL2 | |
Recombinant Protein | |
E. coli | |
QDVRVQVLPE VRGQLGGTVE LPCHLLPPVP GLYISLVTWQ RPDAPANHQN VAAFHPKMGP SFPSPKPGSE RLSFVSAKQS TGQDTEAELQ DATLALHGLT VEDEGNYTCE FATFPKGSVR GMTWLRVIAK PKNQAEAQKV TFSQDPTTVA LCISKEGRPP ARISWLSSLD WEAKETQVSG TLAGTVTVTS RFTLVPSGRA DGVTVTCKVE HESFEEPALI PVTLSVRYPP EVSISGYDDN WYLGRTDATL SCDVRSNPEP TGYDWSTTSG TFPTSAVAQG SQLVIHAVDS LFNTTFVCTV TNAVGMGRAE QVIFVRETPN TAGAGATGG | |
Greater than 90% as determined by SDS-PAGE. |
5819 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |